5-(Trifluoromethyl)-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21858G1 AS21858G25 AS21858G5
ACTIAS21858G1EA 0 GBP
ACTIAS21858G1 ACTIAS21858G25 ACTIAS21858G5
5-(Trifluoromethyl)-1H-indazole
5-(Trifluoromethyl)-1H-indazole
Formula: C₈H₅F₃N₂
MDL Number: MFCD08234904
CAS Number: 885271-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR