1-Acetyl-6-chloro-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6243-5G HC-6243-1G
COBBHC-6243-5GEA 0 GBP
COBBHC-6243-5G COBBHC-6243-1G
1-Acetyl-6-chloro-1H-indazole
1-Acetyl-6-chloro-1H-indazole
Formula: C₉H₇ClN₂O
MDL Number: MFCD09878373
CAS Number: 708-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR