1-Acetyl-4-chloro-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6241-1G HC-6241-5G
COBBHC-6241-1GEA 0 GBP
COBBHC-6241-1G COBBHC-6241-5G
1-Acetyl-4-chloro-1H-indazole
1-Acetyl-4-chloro-1H-indazole
Formula: C₉H₇ClN₂O
MDL Number: MFCD09878372
CAS Number: 145439-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR