6-Bromo-5-methyl-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS51014G1 AS51014G5
ACTIAS51014G1EA 0 GBP
ACTIAS51014G1 ACTIAS51014G5
6-Bromo-5-methyl-1H-indazole
6-Bromo-5-methyl-1H-indazole
Formula: C₈H₇BrN₂
MDL Number: MFCD09026997
CAS Number: 1000343-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR