6-Bromo-3-methyl-1H-indazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1585-1G
HI-1585-5G
COBBHI-1585-1GEA
0
GBP
COBBHI-1585-1G
COBBHI-1585-5G
6-Bromo-3-methyl-1H-indazole
6-Bromo-3-methyl-1H-indazole
Formula:
C₈H₇BrN₂ |
MDL Number:
MFCD08272244 CAS Number: 7746-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...