6-Bromo-3-methyl-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12283G1 AS12283G5
ACTIAS12283G1EA 45 GBP
ACTIAS12283G1 ACTIAS12283G5
6-Bromo-3-methyl-1H-indazole
6-Bromo-3-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD08272244
CAS Number: 7746-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR