6-Bromo-1-methyl-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

IN-0189-5G IN-0189-1G
COBBIN-0189-5GEA 0 GBP
COBBIN-0189-5G COBBIN-0189-1G
6-Bromo-1-methyl-1H-indazole
6-Bromo-1-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD09878569
CAS Number: 590417-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR