6-Bromo-1-methyl-1H-indazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS1902G1
AS1902G5
ACTIAS1902G1EA
0
GBP
ACTIAS1902G1
ACTIAS1902G5
6-Bromo-1-methyl-1H-indazole
6-Bromo-1-methyl-1H-indazole
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol |
MDL Number:
MFCD09878569 CAS Number: 590417-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...