5-Bromo-7-methyl-1H-indazole ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H26753.MD H26753.03
H26753.MDEA 60 GBP
H26753.MD H26753.03
5-Bromo-7-methyl-1H-indazole ≥97%
5-Bromo-7-methyl-1H-indazole

Purity: 97%
CAS No.: 156454-43-2
Molecular Formula: C8H7BrN2
Formula Weight: 211.06
Melting Point: 183-186°
MDL No.: MFCD03990484

Formula: C₈H₇BrN₂
MDL Number: MFCD03990484
CAS Number: 156454-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR