4-Bromo-2-methyl-2H-indazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IN-0205-1G
IN-0205-5G
COBBIN-0205-1GEA
0
GBP
COBBIN-0205-1G
COBBIN-0205-5G
4-Bromo-2-methyl-2H-indazole
4-Bromo-2-methyl-2H-indazole
Formula:
C₈H₇BrN₂ MW: 211.0586 g/mol |
MDL Number:
MFCD09870046 CAS Number: 590417-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...