2-Methyl-2H-indazole-6-boronic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS12298G1
AS12298G5
ACTIAS12298G1EA
0
GBP
ACTIAS12298G1
ACTIAS12298G5
2-Methyl-2H-indazole-6-boronic acid
2-Methyl-2H-indazole-6-boronic acid
Formula:
C₈H₉BN₂O₂ |
MDL Number:
MFCD09870057 CAS Number: 1001907-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...