2-Methyl-2H-indazole-4-boronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

PN-9133-5G PN-9133-1G
COBBPN-9133-5GEA 0 GBP
COBBPN-9133-5G COBBPN-9133-1G
2-Methyl-2H-indazole-4-boronic acid pinacol ester
2-Methyl-2H-indazole-4-boronic acid pinacol ester
Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD11109437
CAS Number: 885698-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR