1-Methyl-1H-indazole-5-boronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methylindazole-5-boronic acid pinacol ester, 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

AS12270G1 AS12270G5
ACTIAS12270G1EA 0 GBP
ACTIAS12270G1 ACTIAS12270G5
1-Methyl-1H-indazole-5-boronic acid pinacol ester
1-Methyl-1H-indazole-5-boronic acid pinacol ester
Formula: C₁₄H₁₉BN₂O₂
MW: 258.123 g/mol
MDL Number: MFCD15071438
CAS Number: 1235469-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR