1-Methyl-1H-indazole-4-boronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

AS12257G1 AS12257G5
ACTIAS12257G1EA 0 GBP
ACTIAS12257G1 ACTIAS12257G5
1-Methyl-1H-indazole-4-boronic acid pinacol ester
1-Methyl-1H-indazole-4-boronic acid pinacol ester
Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD10700158
CAS Number: 885698-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR