4-(4-Bromo-1H-pyrazol-1-yl)-N-dimethylbenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-1119-5G
OT-1119-1G
COBBOT-1119-5GEA
0
GBP
COBBOT-1119-5G
COBBOT-1119-1G
4-(4-Bromo-1H-pyrazol-1-yl)-N-dimethylbenzenesulphonamide
4-(4-Bromo-1H-pyrazol-1-yl)-N-dimethylbenzenesulphonamide
Formula:
C₁₁H₁₂BrN₃O₂S |
MDL Number:
MFCD13195748 CAS Number: 1199773-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...