4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-[5-(4-Dimethylaminophenyl)-2-oxazolyl]benzeneboronic acid

BB-3485-1G BB-3485-250M
COBBBB-3485-1GEA 0 GBP
COBBBB-3485-1G COBBBB-3485-250M
4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid
4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid
Formula: C₁₇H₁₇BN₂O₃
MDL Number: MFCD08689479
CAS Number: 380499-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR