N-(Furan-2-ylmethyl) 3-bromo-4-fluorobenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4633-5G
CA-4633-25G
CA-4633-1G
COBBCA-4633-5GEA
0
GBP
COBBCA-4633-5G
COBBCA-4633-25G
COBBCA-4633-1G
N-(Furan-2-ylmethyl) 3-bromo-4-fluorobenzamide
N-(Furan-2-ylmethyl) 3-bromo-4-fluorobenzamide
Formula:
C₁₂H₉BrFNO₂ |
MDL Number:
MFCD11053851 CAS Number: 1065074-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...