4-Bromo-N-(furan-2-ylmethyl)-3-methoxybenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4647-5G CA-4647-1G
COBBCA-4647-5GEA 0 GBP
COBBCA-4647-5G COBBCA-4647-1G
4-Bromo-N-(furan-2-ylmethyl)-3-methoxybenzamide
4-Bromo-N-(furan-2-ylmethyl)-3-methoxybenzamide
Formula: C₁₃H₁₂BrNO₃
MDL Number: MFCD11504871
CAS Number: 1072944-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR