6-Nitroimidazo[1,2-a]pyridine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6160-1G HC-6160-25G HC-6160-5G
COBBHC-6160-1GEA 0 GBP
COBBHC-6160-1G COBBHC-6160-25G COBBHC-6160-5G
6-Nitroimidazo[1,2-a]pyridine hydrochloride
6-Nitroimidazo[1,2-a]pyridine hydrochloride
Formula: C₇H₆ClN₃O₂
MDL Number: MFCD09800971
CAS Number: 957120-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR