6-Aminoimidazo[1,2-a]pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Imidazo[1,2-a]pyridin-6-amine

AS21689G1 AS21689G5
ACTIAS21689G1EA 41 GBP
ACTIAS21689G1 ACTIAS21689G5
6-Aminoimidazo[1,2-a]pyridine
6-Aminoimidazo[1,2-a]pyridine
Formula: C₇H₇N₃
MW: 133.1506 g/mol
MDL Number: MFCD06739232
CAS Number: 235106-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR