Methyl imidazo[1,2-a]pyridine-7-carboxylate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12860G1 AS12860G5
ACTIAS12860G1EA 0 GBP
ACTIAS12860G1 ACTIAS12860G5
Methyl imidazo[1,2-a]pyridine-7-carboxylate
Methyl imidazo[1,2-a]pyridine-7-carboxylate
Formula: C₉H₈N₂O₂
MDL Number: MFCD09910350
CAS Number: 86718-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR