8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6163-5G HC-6163-1G
COBBHC-6163-5GEA 0 GBP
COBBHC-6163-5G COBBHC-6163-1G
8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride
8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride
Formula: C₇H₅BrClN₃O₂
MDL Number: MFCD09800973
CAS Number: 957120-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR