Imidazo[1,2-b]pyridazine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-7020-1G HC-7020-5G
COBBHC-7020-1GEA 0 GBP
COBBHC-7020-1G COBBHC-7020-5G
Imidazo[1,2-b]pyridazine hydrochloride
Imidazo[1,2-b]pyridazine hydrochloride
Formula: C₆H₆ClN₃
MDL Number: MFCD11226191
CAS Number: 18087-70-2
EINECS: 431-510-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR