Imidazo[1,2-b]pyridazine-6-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12350G1 AS12350G5
ACTIAS12350G1EA 292 GBP
ACTIAS12350G1 ACTIAS12350G5
Imidazo[1,2-b]pyridazine-6-carboxylic acid
Imidazo[1,2-b]pyridazine-6-carboxylic acid
Formula: C₇H₅N₃O₂
MDL Number: MFCD11044772
CAS Number: 769109-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR