Ammonium nitrate-[¹⁵N₂] (99% ¹⁵N), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

366528-1G 366528-250MG
SIAL366528-1GEA 275 GBP
SIAL366528-1G SIAL366528-250MG
Ammonium nitrate-[¹⁵N₂] (99% ¹⁵N), Sigma-Aldrich®
Ammonium nitrate-[15N2]
Formula: H₄N₂O₃
MDL Number: MFCD00082562
CAS Number: 43086-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR