1,1'-Bis(diisopropylphosphino)ferrocene ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: DiPPF, 1,1'-Bis(di-i-propylphosphino)ferrocene

H27057.03 H27057.06
H27057.03EA 115 GBP
H27057.03 H27057.06
1,1'-Bis(diisopropylphosphino)ferrocene ≥98%
1,1'-Bis(diisopropylphosphino)ferrocene

Purity: 98% CAS No.: 97239-80-0 Molecular Formula: C22H36FeP2 Formula Weight: 418.34 Flash Point: 110°(230°F) Melting Point: 50-52° MDL No.: MFCD02684559

Formula: C₂₂H₂₈FeP₂
MW: 410.25 g/mol
Melting Pt: 50…52 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02684559
CAS Number: 97239-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR