3-Chloro-2-fluoro-5-(trifluoromethyl)benzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005683-1G 005683-5G
FLUO005683-1GEA 0 GBP
FLUO005683-1G FLUO005683-5G
3-Chloro-2-fluoro-5-(trifluoromethyl)benzyl alcohol
3-Chloro-2-fluoro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₅ClF₄O
MDL Number: MFCD01631512
CAS Number: 261763-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR