N-Benzyl-4-bromo-3-methylbenzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4553-5G
CA-4553-1G
COBBCA-4553-5GEA
0
GBP
COBBCA-4553-5G
COBBCA-4553-1G
N-Benzyl-4-bromo-3-methylbenzamide
N-Benzyl-4-bromo-3-methylbenzamide
Formula:
C₁₅H₁₄BrNO |
MDL Number:
MFCD09972121 CAS Number: 1020252-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...