4-Bromo-3-(methylamino)benzotrifluoride hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2871-5G HC-2871-1G
COBBHC-2871-5GEA 0 GBP
COBBHC-2871-5G COBBHC-2871-1G
4-Bromo-3-(methylamino)benzotrifluoride hydrochloride
4-Bromo-3-(methylamino)benzotrifluoride hydrochloride
Formula: C₈H₈BrClF₃N
MDL Number: MFCD12913959
CAS Number: 1187386-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR