3-(Methylsulphonyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Methanesulphonyl)phenylboronic acid, 3-(Methylsulphonyl)benzeneboronic acid, (3-(Methylsulphonyl)phenyl)boronic acid

BB-2626-5G BB-2626-25G BB-2626-1G
COBBBB-2626-5GEA 0 GBP
COBBBB-2626-5G COBBBB-2626-25G COBBBB-2626-1G
3-(Methylsulphonyl)phenylboronic acid
3-(Methylsulphonyl)phenylboronic acid
Formula: C₇H₉BO₄S
MW: 200.02 g/mol
MDL Number: MFCD03092935
CAS Number: 373384-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR