8-Bromo-6-methyl-3-nitroimidazo[1,2-a]pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2127-5G HC-2127-1G
COBBHC-2127-5GEA 0 GBP
COBBHC-2127-5G COBBHC-2127-1G
8-Bromo-6-methyl-3-nitroimidazo[1,2-a]pyridine
8-Bromo-6-methyl-3-nitroimidazo[1,2-a]pyridine
Formula: C₈H₆BrN₃O₂
MDL Number: MFCD11504901
CAS Number: 1072944-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR