8-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6161-5G
HC-6161-1G
COBBHC-6161-5GEA
0
GBP
COBBHC-6161-5G
COBBHC-6161-1G
8-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride
8-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride
Formula:
C₈H₈BrClN₂ |
MDL Number:
MFCD09800972 CAS Number: 957120-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...