N-tert-Butylisonicotinamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-(tert-Butyl)isonicotinamide
PY-1295-5G
PY-1295-1G
COBBPY-1295-5GEA
0
GBP
COBBPY-1295-5G
COBBPY-1295-1G
N-tert-Butylisonicotinamide
N-tert-Butylisonicotinamide
Formula:
C₁₀H₁₄N₂O |
MDL Number:
MFCD00466497 CAS Number: 65321-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...