(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4016023.0001
4016023.0005
BACE4016023.0001EA
0
GBP
BACE4016023.0001
BACE4016023.0005
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Formula:
C₁₀H₁₁NO₂ |
MDL Number:
MFCD00144038 CAS Number: 103733-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...