1-Boc-3-piperidineacetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Boc-Piperidine-3-acetic acid

AS20386G1 AS20386G5
ACTIAS20386G1EA 16 GBP
ACTIAS20386G1 ACTIAS20386G5
1-Boc-3-piperidineacetic acid
1-Boc-3-piperidineacetic acid
Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
MDL Number: MFCD02178999
CAS Number: 183483-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR