1-Boc-2-piperidineacetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Boc-2-Piperidineacetic acid

AS22285G1 AS22285G5
ACTIAS22285G1EA 31 GBP
ACTIAS22285G1 ACTIAS22285G5
1-Boc-2-piperidineacetic acid
1-Boc-2-piperidineacetic acid
Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
MDL Number: MFCD02179000
CAS Number: 149518-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR