Borane-tert-butylamine complex, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Boron hydride-tert-butylamine
M-Clarity™ quality level = MQ100
180211-100G
180211-25G
SIAL180211-100GEA
280
GBP
SIAL180211-100G
SIAL180211-25G
Borane-tert-butylamine complex, Sigma-Aldrich®
Borane tert-butylamine complex
Formula:
C₄H₁₄BN MW: 86.98 g/mol |
MDL Number:
MFCD00075635 CAS Number: 7337-45-3 EINECS: 230-851-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...