4-(Butylcarbamoyl)-3-fluorophenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-3313-1G
BB-3313-5G
COBBBB-3313-1GEA
0
GBP
COBBBB-3313-1G
COBBBB-3313-5G
4-(Butylcarbamoyl)-3-fluorophenylboronic acid
4-(Butylcarbamoyl)-3-fluorophenylboronic acid
Formula:
C₁₁H₁₅BFNO₃ |
MDL Number:
MFCD08689527 CAS Number: 874289-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...