4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
AS20745G25
AS20745G5
ACTIAS20745G25EA
59
GBP
ACTIAS20745G25
ACTIAS20745G5
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula:
C₁₄H₁₉BO₄ MW: 262.11 g/mol |
MDL Number:
MFCD02179438 CAS Number: 171364-80-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...