3-(Methoxycarbonyl)phenylboronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20744G25 AS20744G5
ACTIAS20744G25EA 89 GBP
ACTIAS20744G25 ACTIAS20744G5
3-(Methoxycarbonyl)phenylboronic acid pinacol ester
3-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MDL Number: MFCD03789258
CAS Number: 480425-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR