Cyclohexylboronic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
556580-10G
556580-5G
SIAL556580-10GEA
77.3
GBP
SIAL556580-10G
SIAL556580-5G
Cyclohexylboronic acid, Sigma-Aldrich®
Cyclohexylboronic acid
Formula:
C₆H₁₃BO₂ MW: 127.9772 g/mol |
MDL Number:
MFCD01311824 CAS Number: 4441-56-9 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...