Cyclohexylboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
011024-5G
011024-1G
FLUO011024-5GEA
0
GBP
FLUO011024-5G
FLUO011024-1G
Cyclohexylboronic acid
Cyclohexylboronic acid
Formula:
C₆H₁₃BO₂ MW: 127.9772 g/mol |
MDL Number:
MFCD01311824 CAS Number: 4441-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...