2-Methyl-5-nitrophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methyl-5-nitrobenzeneboronic acid

BB-2473-5G BB-2473-1G
COBBBB-2473-5GEA 0 GBP
COBBBB-2473-5G COBBBB-2473-1G
2-Methyl-5-nitrophenylboronic acid
2-Methyl-5-nitrophenylboronic acid
Formula: C₇H₈BNO₄
MW: 180.96 g/mol
MDL Number: MFCD00757435
CAS Number: 100960-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR