4-Bromo-N-methylbenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0078-5G
OT-0078-25G
OT-0078-1G
COBBOT-0078-5GEA
0
GBP
COBBOT-0078-5G
COBBOT-0078-25G
COBBOT-0078-1G
4-Bromo-N-methylbenzenesulphonamide
4-Bromo-N-methylbenzenesulphonamide
Formula:
C₇H₈BrNO₂S MW: 250.1100 g/mol |
MDL Number:
MFCD00993162 CAS Number: 703-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...