4-Bromo-N-ethylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0079-1G OT-0079-5G
COBBOT-0079-1GEA 0 GBP
COBBOT-0079-1G COBBOT-0079-5G
4-Bromo-N-ethylbenzenesulphonamide
4-Bromo-N-ethylbenzenesulphonamide
Formula: C₈H₁₀BrNO₂S
MW: 264.1395 g/mol
MDL Number: MFCD01215100
CAS Number: 1984-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR