3-Bromo-N-butylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0144-5G OT-0144-1G
COBBOT-0144-5GEA 0 GBP
COBBOT-0144-5G COBBOT-0144-1G
3-Bromo-N-butylbenzenesulphonamide
3-Bromo-N-butylbenzenesulphonamide
Formula: C₁₀H₁₄BrNO₂S
MDL Number: MFCD07363822
CAS Number: 871269-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR