5-Bromo-2-methoxy-N-methylbenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0308-5G
OT-0308-1G
COBBOT-0308-5GEA
0
GBP
COBBOT-0308-5G
COBBOT-0308-1G
5-Bromo-2-methoxy-N-methylbenzenesulphonamide
5-Bromo-2-methoxy-N-methylbenzenesulphonamide
Formula:
C₈H₁₀BrNO₃S |
MDL Number:
MFCD06808832 CAS Number: 871269-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...