5-Bromo-N-ethyl-2-methoxybenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0309-5G
OT-0309-1G
COBBOT-0309-5GEA
0
GBP
COBBOT-0309-5G
COBBOT-0309-1G
5-Bromo-N-ethyl-2-methoxybenzenesulphonamide
5-Bromo-N-ethyl-2-methoxybenzenesulphonamide
Formula:
C₉H₁₂BrNO₃S |
MDL Number:
MFCD06003173 CAS Number: 717892-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...