5-Bromo-N-tert-butyl-2-methoxybenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0306-1G
OT-0306-5G
COBBOT-0306-1GEA
0
GBP
COBBOT-0306-1G
COBBOT-0306-5G
5-Bromo-N-tert-butyl-2-methoxybenzenesulphonamide
5-Bromo-N-tert-butyl-2-methoxybenzenesulphonamide
Formula:
C₁₁H₁₆BrNO₃S |
MDL Number:
MFCD08056376 CAS Number: 871269-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...