3-Bromo-N-cyclopropyl-5-nitrobenzamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4375-5G CA-4375-1G
COBBCA-4375-5GEA 0 GBP
COBBCA-4375-5G COBBCA-4375-1G
3-Bromo-N-cyclopropyl-5-nitrobenzamide
3-Bromo-N-cyclopropyl-5-nitrobenzamide
Formula: C₁₀H₉BrN₂O₃
MDL Number: MFCD09475856
CAS Number: 941294-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR